Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylcyclohexanone ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

220435-100G 220435-25G 220435-500G
BT220435-100GEA 123.14 USD InStock
BT220435-100G BT220435-25G BT220435-500G
4-Methylcyclohexanone ≥98%
4-Methylcyclohexanone
CAS: 589-92-4; EC No: 209-665-3; MDL No: MFCD00001643; RTECS: GW1750200 UN No: UN2297; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H12O; MW: 112.17 Boiling Point: 169-171°; Flash point: 48°C (118°F) Density (g/mL): 0.914; Refractive Index: 1.444

  • RTECS: GW1750200
  • Refractive index: 1.444
  • Liquid


Formula: C₇H₁₂O
Boiling Pt: 169…171 °C
Density: 0.914 g/mL
Flash Pt: 48 °C (118 °F)
MDL Number: MFCD00001643
CAS Number: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: 3,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty