Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese(II) sulfate hydrate ≥99.999%

Supplier: BEANTOWN CHEMICAL - BTC

220440-25G 220440-5G
BT220440-25GEA 145 USD InStock
BT220440-25G BT220440-5G
Manganese(II) sulfate hydrate ≥99.999%
Manganese (II) sulphate hydrate
CAS: 15244-36-7; EC No: 232-089-9; MDL No: MFCD00149159 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Powder; Linear Formula: MnSO4·xH2O; MW: 151.00 (anhydrous) Density (g/mL): 2.09 Hygroscopic

  • Hygroscopic
  • Crystalline/Powder


Formula: MnO₄S·H₂O
MW: 169.02 g/mol
Density: 2.09 g/mL
MDL Number: MFCD00149159
CAS Number: 15244-36-7
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty