221140-100G
221140-2.5KG
221140-500G
BT221140-100GEA
26
USD
InStock
BT221140-100G
BT221140-2.5KG
BT221140-500G
N-Methylaniline ≥98%
N-Methylaniline
CAS: 100-61-8; EC No: 202-870-9; MDL No: MFCD00008283; RTECS: BY4550000
UN No: UN2294; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: C6H5NHCH3; Molecular Formula: C7H9N; MW: 107.16
Melting Point: -57°; Boiling Point: 194-197°; Flash point: 85°C (185°F)
Density (g/mL): 0.989; Refractive Index: 1.571
Air Sensitive
- RTECS: BY4550000
- Refractive index: 1.571
- Air Sensitive
- Liquid