Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl thiophosphate

Supplier: BEANTOWN CHEMICAL - BTC

221240-1G 221240-25G 221240-5G
BT221240-1GEA 27 USD InStock
BT221240-1G BT221240-25G BT221240-5G
Trimethyl thiophosphate
Trimethyl thiophosphate
CAS: 152-18-1; MDL No: MFCD00014888 UN No: UN3278; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: P(S)(OCH3)3; Molecular Formula: C3H9O3PS; MW: 156.15

  • Liquid


Formula: C₃H₉O₃PS
MW: 156.136 g/mol
MDL Number: MFCD00014888
CAS Number: 152-18-1
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty