Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron ≥99.9% (trace metals basis), sponge -100 mesh

Supplier: BEANTOWN CHEMICAL - BTC

221865-100G 221865-2KG 221865-500G
BT221865-100GEA 27 USD InStock
BT221865-100G BT221865-2KG BT221865-500G
Iron ≥99.9% (trace metals basis), sponge -100 mesh
Iron
CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999; RTECS: NO4565500 UN No: UN3089; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: Fe; MW: 55.85 Melting Point: 1535°; Boiling Point: 2750° Density (g/mL): 7.86

  • RTECS: NO4565500
  • Solid


Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/mL
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1 III
Merck Index: 13,05109
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty