Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Antimony ≥99.5% (trace metals basis), powder -100 mesh

Supplier: BEANTOWN CHEMICAL - BTC

221885-1KG 221885-250G
BT221885-1KGEA 116 USD InStock
BT221885-1KG BT221885-250G
Antimony ≥99.5% (trace metals basis), powder -100 mesh
Antimony
CAS: 7440-36-0; EC No: 231-146-5; MDL No: MFCD00134030 UN No: UN2871; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: Sb; MW: 121.76 Melting Point: 630°; Boiling Point: 1635° Density (g/mL): 6.69

  • Powder


Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637
Melting Pt: 630
Density: 6.69 g/mL
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty