Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium hydrogenarsenate heptahydrate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

222180-100G 222180-500G
BT222180-100GEA 83.38 USD InStock
BT222180-100G BT222180-500G
Disodium hydrogenarsenate heptahydrate ≥98%
Disodium hydrogenarsenate heptahydrate
CAS: 10048-95-0; EC No: 231-902-4; MDL No: MFCD00149179; RTECS: CG0900000 UN No: UN1685; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Na2HAsO4·7H2O; MW: 312.01 Melting Point: 180° Density (g/mL): 1.88

  • RTECS: CG0900000
  • Crystalline


Formula: HAsNa₂O₄·7H₂O
MW: 312.01 g/mol
Melting Pt: 57
Density: 1.88 g/mL
MDL Number: MFCD00149179
CAS Number: 10048-95-0
EINECS: 231-902-4
UN: 1685
ADR: 6.1 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty