Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl vinylphosphonate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

222360-25G 222360-5G
BT222360-25GEA 147 USD InStock
BT222360-25G BT222360-5G
Dimethyl vinylphosphonate ≥98%
Dimethyl vinylphosphonate
CAS: 4645-32-3; EC No: 225-076-4; MDL No: MFCD00015488 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C4H9O3P; MW: 136.09 Boiling Point: 197-203°; Flash point: 98°C (208°F) Density (g/mL): 1.146; Refractive Index: 1.428 Air Sensitive, Light Sensitive, Moisture Sensitive

  • Refractive index: 1.428
  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Liquid


Formula: C₄H₉O₃P
Boiling Pt: 197…203 °C
Density: 1.146 g/mL
Flash Pt: 98 °C (208 °F)
MDL Number: MFCD00015488
CAS Number: 4645-32-3
EINECS: 225-076-4
ADR: 8,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty