222395-100G
222395-25G
BT222395-100GEA
301.67
USD
InStock
BT222395-100G
BT222395-25G
Aluminium iodide ≥95%
Aluminium iodide
CAS: 7784-23-8; EC No: 232-054-8; MDL No: MFCD00003427
UN No: UN3260; Haz Class: 8; Packing Group: II
Granular; Molecular Formula: AlI3; MW: 407.70
Melting Point: 191°; Boiling Point: 360°
Density (g/mL): 3.98
Light Sensitive, Moisture Sensitive
- Light Sensitive, Moisture Sensitive
- Granular