Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl bromoacetate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

222400-1KG 222400-250G 222400-50G
BT222400-1KGEA 309 USD InStock
BT222400-1KG BT222400-250G BT222400-50G
Methyl bromoacetate ≥98%
Methyl bromoacetate
CAS: 96-32-2; EC No: 202-499-2; MDL No: MFCD00000189; RTECS: AF6300000 UN No: UN2643; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: BrCH2COOCH3; Molecular Formula: C3H5BrO2; MW: 152.98 Melting Point: -50°; Boiling Point: 144-145°; Flash point: 64°C (147°F) Density (g/mL): 1.616; Refractive Index: 1.458

  • RTECS: AF6300000
  • Refractive index: 1.458
  • Liquid


Formula: BrCH₂COOCH₃
Boiling Pt: 144…145 °C
Melting Pt: –50 °C
Density: 1.616 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD00000189
CAS Number: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty