Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylbutanone ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

222460-1L 222460-250ML
BT222460-1LEA 134.72 USD InStock
BT222460-1L BT222460-250ML
3-Methylbutanone ≥98%
3-Methylbutanone
CAS: 563-80-4; EC No: 209-264-3; MDL No: MFCD00008919; RTECS: EL9100000 UN No: UN2397; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHCOCH3; Molecular Formula: C5H10O ; MW: 86.13 Melting Point: -92°; Boiling Point: 94-95°; Flash point: -1°C (30°F) Density (g/mL): 0.805 ; Refractive Index: 1.388

  • RTECS: EL9100000
  • Refractive index: 1.388
  • Liquid


Formula: C₅H₁₀O
Boiling Pt: 94…95 °C
Melting Pt: –92 °C
Density: 0.805 g/mL
Flash Pt: –1 °C (30 °F)
MDL Number: MFCD00008919
CAS Number: 563-80-4
EINECS: 209-264-3
UN: 2397
ADR: 3,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty