Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Triisopropylbenzenesulphonyl chloride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

222535-100G 222535-25G 222535-500G
BT222535-100GEA 182 USD InStock
BT222535-100G BT222535-25G BT222535-500G
2,4,6-Triisopropylbenzenesulphonyl chloride ≥98%
2,4,6-Triisopropylbenzenesulphonyl chloride
CAS: 6553-96-4; EC No: 229-479-6; MDL No: MFCD00007433 UN No: UN3096; Haz Class: 8 (4.3); Packing Group: II Crystalline; Molecular Formula: C15H23ClO2S; MW: 302.86 Melting Point: 92-94° Moisture Sensitive

  • Moisture Sensitive
  • Crystalline


Formula: C₁₅H₂₃ClO₂S
MW: 302.87
Melting Pt: 92…94 °C
MDL Number: MFCD00007433
CAS Number: 6553-96-4
EINECS: 229-479-6
UN: 3096
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty