223120-1G
223120-5G
BT223120-1GEA
184
USD
InStock
BT223120-1G
BT223120-5G
3-Fluoro-4-iodopyridine ≥98%
3-Fluoro-4-iodopyridine
CAS: 22282-75-3; MDL No: MFCD03092927
UN No: UN3261 ; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: C5H3FIN ; MW: 222.99
Melting Point: 85-89°