223530-100G
223530-2.5KG
223530-500G
BT223530-100GEA
36.43
USD
InStock
BT223530-100G
BT223530-2.5KG
BT223530-500G
Nickel acetate tetrahydrate ≥98%
Nickel acetate tetrahydrate
CAS: 6018-89-9; EC No: 206-761-7; MDL No: MFCD00066973; RTECS: QR6126000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: Ni(OCOCH3)2·4H2O; MW: 248.84
Density (g/mL): 1.798
- RTECS: QR6126000
- Crystalline