Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cholesteryl chloroformate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

223925-100G 223925-25G
BT223925-100GEA 105.36 USD InStock
BT223925-100G BT223925-25G
Cholesteryl chloroformate ≥98%
Cholesteryl chloroformate
CAS: 7144-08-3; EC No: 230-447-9; MDL No: MFCD00003633 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline/Powder; Molecular Formula: C28H45ClO2; MW: 449.11 Melting Point: 115-117° Optical Rotation: [α]27/D -28°, c = 2 in chloroform Moisture Sensitive

  • Optical rotation: [α]27/D -28°, c = 2 in chloroform
  • Moisture Sensitive
  • Crystalline/Powder


Formula: C₂₈H₄₅ClO₂
MW: 449.12 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD00003633
CAS Number: 7144-08-3
EINECS: 230-447-9
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty