224245-100G
224245-2.5KG
224245-500G
BT224245-100GEA
41.55
USD
InStock
BT224245-100G
BT224245-2.5KG
BT224245-500G
Sodium chromate, anhydrous ≥98%
Sodium chromate
CAS: 7775-11-3; EC No: 231-889-5; MDL No: MFCD00003484; RTECS: GB2955000
UN No: UN3288; Haz Class: 6.1; Packing Group: II
Solid; Linear Formula: Na2CrO4; MW: 161.97
Melting Point: 792°
Density (g/mL): 2.72
Hygroscopic
- RTECS: GB2955000
- Hygroscopic
- Solid