Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isohexane (mixture of isomeric branched chain Hexanes) environmental

Supplier: BEANTOWN CHEMICAL - BTC

224845-1L 224845-4L
BT224845-1LEA 52 USD InStock
BT224845-1L BT224845-4L
Isohexane (mixture of isomeric branched chain Hexanes) environmental
Isohexane (mixture of isomeric branched chain Hexanes)
CAS: 92112-69-1; EC No: 295-570-2; MDL No: MFCD02179311 UN No: UN1208; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H14 ; MW: 86.18 Melting Point: -95°; Boiling Point: 68-70°; Flash point: -23°C (-9°F) Density (g/mL): 0.672; Refractive Index: 1.379

  • Refractive index: 1.379
  • Liquid


Formula: C₆H₁₄
MDL Number: MFCD02179311
CAS Number: 73513-42-5
EINECS: 295-570-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty