Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-1-Phenylethyl isocyanate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

224975-1G 224975-5G
BT224975-1GEA 46 USD InStock
BT224975-1G BT224975-5G
(S)-(-)-1-Phenylethyl isocyanate ≥98%
(S)-(-)-1-Phenylethyl isocyanate
CAS: 14649-03-7; EC No: 238-698-6; MDL No: MFCD00002036 UN No: UN2206; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C9H9NO; MW: 147.17 Boiling Point: 55-56°/2.5 mmHg; Flash point: 65°C (149°F) Density (g/mL): 1.045; Refractive Index: 1.515; Optical Rotation: [α]20/D -10°, neat Moisture Sensitive

  • Optical rotation: [α]20/D -10°, neat
  • Refractive index: 1.515
  • Moisture Sensitive
  • Liquid


Formula: C₉H₉NO
Boiling Pt: 55…56 °C/2.5 mmHg
Density: 1.045 g/mL
Flash Pt: 65 °C (149 °F)
MDL Number: MFCD00002036
CAS Number: 14649-03-7
EINECS: 238-698-6
ADR: 6.1,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty