Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsilyl cyanide ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

225530-100G 225530-25G 225530-5G
BT225530-100GEA 425 USD InStock
BT225530-100G BT225530-25G BT225530-5G
Trimethylsilyl cyanide ≥97%
Trimethylsilyl cyanide
CAS: 7677-24-9; EC No: 231-657-3; MDL No: MFCD00001765 UN No: UN3383; Haz Class: 6.1 (3); Packing Group: I Liquid; Linear Formula: Si(CH3)3CN; Molecular Formula: C4H9NSi; MW: 99.21 Melting Point: 8-11°; Boiling Point: 114-117°; Flash point: 1°C (34°F) Density (g/mL): 0.793; Refractive Index: 1.392 Moisture Sensitive

  • Refractive index: 1.392
  • Moisture Sensitive
  • Liquid


Formula: C₄H₉NSi
MW: 99.21 g/mol
Boiling Pt: 114
Melting Pt: 11
Density: 0.793 g/mL
Flash Pt: 1 °C (34 °F)
MDL Number: MFCD00001765
CAS Number: 7677-24-9
EINECS: 231-657-3
UN: 3383
ADR: 6.1 I
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty