Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dinitrobenzenesulphonyl chloride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

225975-25G 225975-5G
BT225975-25GEA 359 USD InStock
BT225975-25G BT225975-5G
2,4-Dinitrobenzenesulphonyl chloride ≥98%
2,4-Dinitrobenzenesulphonyl chloride
CAS: 1656-44-6; EC No: 216-749-3; MDL No: MFCD00007431 UN No: UN3096; Haz Class: 8 (4.3); Packing Group: II Crystalline; Molecular Formula: C6H3ClN2O6S ; MW: 266.62 Melting Point: 101-103° Moisture Sensitive

  • Moisture Sensitive
  • Crystalline


Formula: C₆H₃ClN₂O₆S
MW: 266.62 g/mol
Melting Pt: 102
MDL Number: MFCD00007431
CAS Number: 1656-44-6
EINECS: 216-749-3
UN: 3096
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty