Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1-Adamantyl)-4-methylphenol 99%

Supplier: Origen Biomedical, Inc
Warning

H55922-03 H55922-06
AAH55922-03EA 75.9 USD InStock
AAH55922-03 AAH55922-06 AAH55922-MD
2-(1-Adamantyl)-4-methylphenol 99%
2-(1-Adamantyl)-4-methylphenol
Purity: 99% CAS No.: 41031-50-9 Molecular Formula: C17H22O Formula Weight: 242.36 Physical Form: Powder Appearance: White Melting Point: 128-130° MDL No.: MFCD00168147


Formula: C₁₇H₂₂O
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00168147
CAS Number: 41031-50-9
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty