Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-His-OMe 95%

Supplier: Origen Biomedical, Inc

H62545-03 H62545-06
AAH62545-03EA 83.3 USD InStock
AAH62545-03 AAH62545-06 AAH62545-MD
Boc-His-OMe 95%
Boc-His-OMe
Purity: 95% CAS No.: 2488-14-4 Molecular Formula: C12H19N3O4 Formula Weight: 269.30 Physical Form: Solid MDL No.: MFCD00167635


Formula: C₁₂H₁₉N₃O₄
MW: 269.301 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167635
CAS Number: 2488-14-4
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty