Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tritylthio)propanoate 98%

Supplier: Origen Biomedical, Inc

H63003-03 H63003-06
AAH63003-03EA 65.1 USD InStock
AAH63003-03 AAH63003-06 AAH63003-MD
(R)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tritylthio)propanoate 98%
(R)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tritylthio)propanoate
Purity: 98% CAS No.: 115520-21-3 Molecular Formula: C43H30F5NO4S Formula Weight: 751.76 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 4225739 MDL No.: MFCD00065639


Formula: C₄₃H₃₀F₅NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00065639
CAS Number: 115520-21-3
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty