Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzoyl-L-tyrosine ethyl ester 98%

Supplier: Origen Biomedical, Inc

H63434-06 H63434-14
AAH63434-06EA 38.9 USD InStock
AAH63434-06 AAH63434-14
N-Benzoyl-L-tyrosine ethyl ester 98%
N-Benzoyl-L-tyrosine ethyl ester
Purity: 98% CAS No.: 3483-82-7 Molecular Formula: C18H19NO4 Formula Weight: 313.35 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 118-121° Beilstein Reference: 2223819 MDL No.: MFCD00002388


Formula: C₁₈H₁₉NO₄
Melting Pt: 118…121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002388
CAS Number: 3483-82-7
EINECS: 222-469-2
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty