Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiazole-4-carboxaldehyde 95%

Supplier: Origen Biomedical, Inc
Warning

H64028-03 H64028-06 H64028-MD
AAH64028-03EA 80.4 USD InStock
AAH64028-03 AAH64028-06 AAH64028-MD
Thiazole-4-carboxaldehyde 95%
Thiazole-4-carboxaldehyde
Purity: 95% CAS No.: 3364-80-5 Molecular Formula: C4H3NOS Formula Weight: 113.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Pale brown Melting Point: 61-65° Beilstein Reference: 107137 MDL No.: MFCD00626896


Formula: C4H3NOS
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00626896
CAS Number: 3364-80-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty