H64045-14
H64045-22
AAH64045-14EA
56
USD
InStock
AAH64045-14
AAH64045-22
1-Bromo-2-fluoro-5-nitrobenzene 95%
1-Bromo-2-fluoro-5-nitrobenzene
Purity: 95%
CAS No.: 701-45-1
Molecular Formula: C6H3BrFNO2
Formula Weight: 220.00
Physical Form: Solid
Beilstein Reference: 2089367
MDL No.: MFCD00055433