Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-nitropyridine 98%

Supplier: Origen Biomedical, Inc
Warning

H64210-06 H64210-14
AAH64210-06EA 54 USD InStock
AAH64210-06 AAH64210-14
2-Fluoro-5-nitropyridine 98%
2-Fluoro-5-nitropyridine
Purity: 98% CAS No.: 456-24-6 Molecular Formula: C5H3FN2O2 Formula Weight: 142.09 Physical Form: Liquid Density: 1.41 Refractive Index: 1.5250 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Melting Point: 17-21° Boiling Point: 86-87°/7mm lit. Beilstein Reference: 120351 MDL No.: MFCD03095059


Formula: C₅H₃FN₂O₂
MW: 142.09
Boiling Pt: 86…87 °C/7 mm (lit.)
Melting Pt: 17…21 °C
Density: 1.41
Storage Temperature: Ambient
MDL Number: MFCD03095059
CAS Number: 456-24-6
EINECS: 207-260-6
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty