Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-fluoropyridine 98%

Supplier: Origen Biomedical, Inc
Warning

H64233-03 H64233-06
AAH64233-03EA 124 USD InStock
AAH64233-03 AAH64233-06 AAH64233-MD
2-Amino-4-fluoropyridine 98%
2-Amino-4-fluoropyridine
Purity: 98% CAS No.: 944401-77-8 Molecular Formula: C5H5FN2 Formula Weight: 112.04 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Pale yellow Beilstein Reference: 15519014 MDL No.: MFCD09260906


Formula: C₅H₅FN₂
MW: 112.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260906
CAS Number: 944401-77-8
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty