Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-chloro-3-nitropyridine 95%

Supplier: Origen Biomedical, Inc
Warning

H64678-03 H64678-06 H64678-MD
AAH64678-03EA 84.4 USD InStock
AAH64678-03 AAH64678-06 AAH64678-MD
2-Amino-4-chloro-3-nitropyridine 95%
2-Amino-4-chloro-3-nitropyridine
Purity: 95% CAS No.: 6980-08-1 Molecular Formula: C5H4ClN3O2 Formula Weight: 173.56 Beilstein Reference: 391939 MDL No.: MFCD04116097


Formula: C₅H₄ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04116097
CAS Number: 6980-08-1
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty