Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromophthalazin-1(4H)-one 98%

Supplier: Origen Biomedical, Inc
Warning

H65609-03 H65609-06
AAH65609-03EA 65.3 USD InStock
AAH65609-03 AAH65609-06 AAH65609-MD
6-Bromophthalazin-1(4H)-one 98%
6-Bromophthalazin-1(4H)-one
Purity: 98% CAS No.: 75884-70-7 Molecular Formula: C8H5BrN2O Formula Weight: 225.04 Physical Form: Solid Beilstein Reference: 10426132 MDL No.: MFCD09264003


Formula: C₈H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09264003
CAS Number: 75884-70-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty