H65996-03
H65996-MD
AAH65996-03EA
413
USD
InStock
AAH65996-03
AAH65996-MD
2,5-Dioxopyrrolidin-1-yl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate 95%
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester)
Purity: 95%
CAS No.: 55750-63-5
Molecular Formula: C14H16N2O6
Formula Weight: 308.29
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light and Moisture Sensitive; Hygroscopic
Appearance: Off-white to pale brown
Melting Point: 69-74°
Beilstein Reference: 1499815
MDL No.: MFCD00043043