Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-6-methylpyridine 98%

Supplier: Origen Biomedical, Inc
Warning

H65997-03 H65997-06
AAH65997-03EA 89.5 USD InStock
AAH65997-03 AAH65997-06 AAH65997-MD
2-Acetyl-6-methylpyridine 98%
2-Acetyl-6-methylpyridine
Purity: 98% CAS No.: 6940-57-4 Molecular Formula: C8H9NO Formula Weight: 135.17 Storage Temperature: 2°C to 8°C Flash Point: 84°(183°F) Physical Form: Solid Refractive Index: 1.5150 Appearance: Pale yellow Boiling Point: 209° Beilstein Reference: 109564 MDL No.: MFCD06658293


Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 209 °C
Flash Pt: 84 °C(183 °F)
Storage Temperature: Ambient
MDL Number: MFCD06658293
CAS Number: 6940-57-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty