H66396-03
H66396-06
AAH66396-03EA
42.2
USD
InStock
AAH66396-03
AAH66396-06
5-Bromoindole-2-carboxylic acid 96%
5-Bromoindole-2-carboxylic acid
Purity: 96%
CAS No.: 7254-19-5
Molecular Formula: C9H6BrNO2
Formula Weight: 240.06
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 153253
MDL No.: MFCD00022705