Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-methoxy-1H-indole-2-carboxylate 97%

Supplier: Origen Biomedical, Inc
Warning

H66528-03 H66528-06
AAH66528-03EA 97.8 USD InStock
AAH66528-03 AAH66528-06
Methyl-6-methoxy-1H-indole-2-carboxylate 97%
Methyl-6-methoxy-1H-indole-2-carboxylate
Purity: 97% CAS No.: 98081-83-5 Molecular Formula: C11H11NO3 Formula Weight: 205.21 Physical Form: Solid Melting Point: 117-119° Beilstein Reference: 395862 MDL No.: MFCD00134301


Formula: C₁₁H₁₁NO₃
MW: 205.213 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00134301
CAS Number: 98081-83-5
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty