H66602-03
H66602-MD
AAH66602-03EA
365
USD
InStock
AAH66602-03
AAH66602-MD
4-(Piperazin-1-yl)-1H-indole 95%
4-(Piperazin-1-yl)-1H-indole
Purity: 95%
CAS No.: 84807-09-0
Molecular Formula: C12H15N3
Formula Weight: 201.27
Physical Form: Solid
Beilstein Reference: 9198621
MDL No.: MFCD00831776