H66850-03
H66850-MD
AAH66850-03EA
129
USD
InStock
AAH66850-03
AAH66850-MD
7-Hydroxy-1-indanone 97%
7-Hydroxy-1-indanone
Purity: 97%
CAS No.: 6968-35-0
Molecular Formula: C9H8O2
Formula Weight: 148.16
Physical Form: Solid
Melting Point: 109-113°
Beilstein Reference: 11085807
MDL No.: MFCD06659676