Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bialaphos sodium ≥85%

Supplier: Origen Biomedical, Inc
Danger

J66802-MC J66802-ME
AAJ66802-MCEA 265.32 USD InStock
AAJ66802-MC AAJ66802-ME
Bialaphos sodium ≥85%
Bialaphos sodium
Has successfully been used in major cereal species transformation

Pale orange powder

Soluble in water


Formula: C₁₁H₂₁N₃NaO₆P
Boiling Pt: 793 °C
Melting Pt: 160 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01674432
CAS Number: 71048-99-2
UN: 2811
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 85

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty