Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Coelenterazine (native) , Aequorea sp.

Supplier: Origen Biomedical, Inc

J66823-MA J66823-MCR
AAJ66823-MAEA 468 USD InStock
AAJ66823-MA AAJ66823-MCR
Coelenterazine (native) , Aequorea sp.
Coelenterazine
A sensitive and specific chemiluminescence probe of the superoxide anion

CAS No.: 55779-48-1 Molecular Formula: C26H21N3O3 Formula Weight: 423.46 Storage Temperature: -30°C to -10°C Physical Form: Solid Sensitivity: Moisture and Light Sensitive Appearance: Yellow to dark orange Solubility: Soluble in methanol or ethanol MDL No.: MFCD00467176

Applications: A sensitive and specific chemiluminescence probe of the superoxide anion


Formula: C₂₆H₂₁N₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00467176
CAS Number: 55779-48-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty