Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycine, 0.2 M Buffer Solution, pH 2.5, Low Endotoxin, Thermo Scientific

Supplier: Origen Biomedical, Inc

J67094-AE J67094-AK
AAJ67094-AEEA 86.08 USD InStock
AAJ67094-AE AAJ67094-AK
Glycine, 0.2 M Buffer Solution, pH 2.5, Low Endotoxin, Thermo Scientific
Buffers Biological Buffers
Liquid

CAS No.: 56-40-6
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00008131


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty