Packaged under argon in resealable ChemSeal™ bottles.
047104-AK
047104-K2
AA47104-AKEA
63.62
USD
InStock
AA47104-AK
AA47104-K2
n-Hexane, anhydrous, over molecular sieves
n-Hexane
CAS No.: 110-54-3
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -23°(-9°F)
Physical Form: Liquid
Density: 0.659
Refractive Index: 1.3750
Appearance: Colorless
Melting Point: -95°
Boiling Point: 69°
Beilstein Reference: 1730733
UN No.: 1208
MDL No.: MFCD00009520