Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Toluene, anhydrous ≥99.8%, over molecular sieves

Supplier: Origen Biomedical, Inc
Danger

Packaged under argon in resealable ChemSeal™ bottles.

047136-AE 047136-K2
AA47136-AEEA 47.99 USD InStock
AA47136-AE AA47136-K2
Toluene, anhydrous ≥99.8%, over molecular sieves
Toluene
Purity: 99.8% CAS No.: 108-88-3 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: 4°(40°F) Physical Form: Liquid Density: 0.867 Refractive Index: 1.4967 Appearance: Colorless Odor: Petroleum-like Melting Point: -93° Boiling Point: 111° Beilstein Reference: 635760 UN No.: 1294 MDL No.: MFCD00008512


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867
Flash Pt: 4 °C (40 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 14,9529
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=7b8a82c36ede4fc1e8822a573f5ce08743569a24&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Minimum 99.8

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty