Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl blue

Supplier: Origen Biomedical, Inc
Warning

H37721-14 H37721-22
AAH37721-14EA 65.92 USD InStock
AAH37721-14 AAH37721-22
Methyl blue
Methyl blue
Methyl blue

CAS No.: 28983-56-4 Molecular Formula: C37H27N3Na2O9S3 Formula Weight: 799.80 Physical Form: Powder Appearance: Blue Odor: Odorless MDL No.: MFCD00058509


Formula: C₃₇H₂₇N₃Na₂O₉S₃
MW: 799.81
Storage Temperature: Ambient
MDL Number: MFCD00058509
CAS Number: 28983-56-4
EINECS: 249-352-9
Merck Index: 13,06055
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty