H64067-03
H64067-06
AAH64067-03EA
137
USD
InStock
AAH64067-03
AAH64067-06
AAH64067-MD
2,2-dimethylbut-3-enoic acid 95%
2,2-dimethylbut-3-enoic acid
2,2-Dimethyl-3-butenoic acid, 95%
Purity: 95%
CAS No.: 10276-09-2
Molecular Formula: C6H10O2
Formula Weight: 114.14
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 0.957
Refractive Index: 1.4295
Appearance: Colorless
Melting Point: -6°
Boiling Point: 185°
Beilstein Reference: 1700758
UN No.: 3265
MDL No.: MFCD00196967