H66219-14
H66219-22
AAH66219-14EA
173
USD
InStock
AAH66219-14
AAH66219-22
2-Amino-1-cyclopentene-1-carbonitrile 98%
2-Amino-1-cyclopentene-1-carbonitrile
2-Amino-1-cyclopentene-1-carbonitrile, 98%
Purity: 98%
CAS No.: 2941-23-3
Molecular Formula: C6H8N2
Formula Weight: 108.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
Melting Point: 147-148°
Beilstein Reference: 742061
UN No.: 3439
MDL No.: MFCD00517551