Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Trp-OMe 96%

Supplier: Origen Biomedical, Inc

H66442-06 H66442-14
AAH66442-06EA 50.6 USD InStock
AAH66442-06 AAH66442-14 AAH66442-22
Boc-Trp-OMe 96%
Boc-Trp-OMe
N-Boc-L-tryptophan methyl ester, 96%

Purity: 96% CAS No.: 33900-28-6 Molecular Formula: C17H22N2O4 Formula Weight: 318.37 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white MDL No.: MFCD01075094


Formula: C₁₇H₂₂N₂O₄
MW: 318.373 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01075094
CAS Number: 33900-28-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty