H66760-03
H66760-MD
AAH66760-03EA
684
USD
InStock
AAH66760-03
AAH66760-MD
4-Amino-2-(chloromethyl)pyrimidine 96%
4-Amino-2-(chloromethyl)pyrimidine
4-Amino-2-(chloromethyl)pyrimidine, 96%
Purity: 96%
CAS No.: 79651-35-7
Molecular Formula: C5H6ClN3
Formula Weight: 143.57
Physical Form: Liquid
Density: 1.35
Appearance: Colorless to yellow
Boiling Point: 264°
MDL No.: MFCD10697353