H66868-06
H66868-14
AAH66868-06EA
69.9
USD
InStock
AAH66868-06
AAH66868-14
N-(tert-Butoxycarbonyl)iminodiacetic acid 98%
N-(tert-Butoxycarbonyl)iminodiacetic acid
N-Boc-iminodiacetic acid, 98%
Purity: 98%
CAS No.: 56074-20-5
Molecular Formula: C9H15NO6
Formula Weight: 233.22
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4435233
MDL No.: MFCD00800351