Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester 96%

Supplier: Origen Biomedical, Inc

H62234-03 H62234-MD
AAH62234-03EA 602 USD InStock
AAH62234-03 AAH62234-MD
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester 96%
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester
3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%

Purity: 96% Molecular Formula: C12H16BFO3 Formula Weight: 238.06 Physical Form: Solid


Formula: C₁₂H₁₆BFO₃
MW: 238.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994409
CAS Number: 1887126-77-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty