Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester 96%

Supplier: Origen Biomedical, Inc

H62587-03 H62587-MD
AAH62587-03EA 624 USD InStock
AAH62587-03 AAH62587-MD
2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester 96%
2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester
2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester, 96%

Purity: 96% Molecular Formula: C13H16BFO4 Formula Weight: 266.07 Physical Form: Solid


Formula: C₁₃H₁₆BFO₄
MW: 266.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23098820
CAS Number: 1429340-48-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty