Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-nitrophenylboronic acid pinacol ester 96%

Supplier: Origen Biomedical, Inc

H62718-03 H62718-MD
AAH62718-03EA 364 USD InStock
AAH62718-03 AAH62718-MD
2-Fluoro-4-nitrophenylboronic acid pinacol ester 96%
2-Fluoro-4-nitrophenylboronic acid pinacol ester


Formula: C₁₂H₁₅BFNO₄
MW: 267.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264075
CAS Number: 1073353-89-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty